Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5029

Sigma-Aldrich

FAK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FADK, FAK1, FRNK, PTK2, pp125FAK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

24-32 nmol/min·mg

mol wt

~140 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTK2(5747)

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FAK signaling in anaplastic astrocytoma and glioblastoma tumors.
Natarajan M
Cancer (2003)
Phosphatidylinositol 4,5-bisphosphate triggers activation of focal adhesion kinase by inducing clustering and conformational changes.
Gorii GM
Proceedings of the National Academy of Sciences of the USA (2014)
Amplification of F-Actin Disassembly and Cellular Repulsion by Growth Factor Signaling
Jimok Yoon
Developmental Cell (2017)
Focal adhesion kinase is involved in rabies virus infection through its interaction with viral phosphoprotein P.
Fouquet B
Journal of Virology (2015)
Gain of miR-151 on chromosome 8q24.3 facilitates
tumour cell migration and spreading through
downregulating RhoGDIA
Nature Cell Biology (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service