Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5022

Sigma-Aldrich

EEF2K, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HSU93850, MGC45041, eEF-2K

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

42-58 nmol/min·mg

mol wt

~125 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EEF2K(29904)

General description

EEF2K is a highly conserved calmodulin-dependent protein kinase that links activation of cell surface receptors to cell division. EEF2K is involved in the regulation of protein synthesis. It phosphorylates eukaryotic elongation factor 2 (EEF2), an abundant cytoplasmic protein that catalyzes the movement of the ribosome along mRNA during translation in eukaryotic cells, and inhibits the EEF2 function. EEF2K is highly expressed in heart and skeletal muscle, suggesting that EEF2 phosphorylation may be particularly important in muscle. EEF2K is highly expressed in patients with systemic lupus erythematosus as well as in many cancers.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A G Ryazanov et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(10), 4884-4889 (1997-05-13)
The several hundred members of the eukaryotic protein kinase superfamily characterized to date share a similar catalytic domain structure, consisting of 12 conserved subdomains. Here we report the existence and wide occurrence in eukaryotes of a protein kinase with a
Sonia Arora et al.
Biochemical and biophysical research communications, 293(3), 1073-1076 (2002-06-08)
Elongation factor 2 kinase (eEF-2K), also known as calmodulin-dependent protein kinase III, is a member of the calmodulin-mediated signaling pathway that links activation of cell surface receptors to cell division. The activity of eEF-2K is increased in many human cancers

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service