Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4966

Sigma-Aldrich

VEGI/TNFSF-15 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

TNFSF15, VEGI isoform, VEGI192A, Vascular Endothelial Growth Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~21.8 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

VEGI (Vascular Endothelial Growth Inhibitor) is a member of the tumor necrosis factor family. VEGI is predominantly an endothelial cell-specific gene, and recombinant VEGI is a potent inhibitor of endothelial cell proliferation, angiogenesis and tumor growth. VEGI exerts two activities of endothelial cells: early Gl arrest of G0/G1-cells responding to growth stimuli, and programmed cell death. VEGI is also able to regulate the expression of several important genes involved in angiogenesis. Human VEGF is a 21.8 kDa homodimeric protein consisting of 192 amino acid polypeptide chains.

Physical form

Lyophilized from a solution containing 0.5 M NaCl, 50 mM Tris-HCl, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1–1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service