Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4964

Sigma-Aldrich

VEGF121 murine

recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)

Synonym(s):

Vegf, Vegf120, Vegf164, Vegf188, Vegfa, Vpf

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Product SRP4964 is not currently sold in your country. Contact Technical Service

biological source

rat

recombinant

expressed in E. coli

assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

potency

1-5 ng/mL

mol wt

~28.2 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VEGFA(7422)

General description

Vascular endothelial growth factor 121 (VEGF121) belongs to the platelet-derived growth factor (PDGF)/VEGF family characterized by the presence of eight conserved cysteine residues and a cystine knot structure.[1] VEGF121 is a disulfide-linked homodimer containing 121 amino acids and is the only variant of VEGF that does not interact with heparin.[2]
Recombinant Mouse VEGF-121 is a truncated version of Mouse VEGF-165.

Biochem/physiol Actions

VEGF121/rGel (toxin gelonin) fusion plays a vital role in targeted destruction of tumor vasculature in vivo and it can be used as a potent anti-cancer agent in cancer treatment. In mice, overexpression of VEGF121 enhanced the tumor growth, whereas VEGF165 decreased tumor growth, without affecting cancer cell proliferation in culture.[2]

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute the lyophilized product with sterile water at a concentration of 0.1 – 0.5 mg/mL, which can be further diluted into other aqueous solutions.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The splice variants of vascular endothelial growth factor (VEGF) and their receptors.
Robinson CJ and Stringer SE.
Journal of Cell Science, 114(pt5), 853-865 (2001)
In vitro and in vivo studies of a VEGF121/rGelonin chimeric fusion toxin targeting the neovasculature of solid tumors.
Veenendaal LM
Proceedings of the National Academy of Sciences of the USA, 99(12), 7866-7871 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service