Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4887

Sigma-Aldrich

MIA human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CD-RAP, Cartilage-derived retinoic acid-sensitive protein, MIA, Melanoma-derived growth regulatory protein precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~12.2 kDa

packaging

pkg of 20 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

human ... MIA(8190)

General description

MIA is the first discovered member of a family of secreted cytokines termed the MIA/OTOR family. The four known members of this family; MIA, MIA2, OTOR and TANGO each contain a Src homology-3 (SH3)-like domain. MIA is an autocrine growth regulatory protein secreted from chondrocytes and malignant melanoma cells that promotes melanoma metastasis by binding competitively to fibronectin and laminin in a manner that results in melanoma cell detachment from the extracellular matrix in vivo. Elevated levels of MIA may represent a clinically useful marker for diagnosis of melanoma metastasis as well as a potential marker for rheumatoid arthritis. Recombinant human MIA is a 12.2 kDa globular protein containing 108 amino acid residues including two intramolecular disulfide bonds.

Physical form

Sterile filtered and lyophilized powder.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. The solution can then be diluted into other aqueous buffers.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumihisa Orita et al.
BMC musculoskeletal disorders, 12, 134-134 (2011-06-18)
The exact mechanism of knee osteoarthritis (OA)-associated pain is unclear, whereas mixed evidence of inflammatory pain and neuropathic pain has been noted. We aimed to investigate pain-related sensory innervation in a monoiodoacetate (MIA)-induced model of OA. Sixty of seventy female

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service