Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4817

Sigma-Aldrich

Keratin-8 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

CK-8, Cytokeratin-8, K8, KRT8, Keratin type II cytoskeletal 8, Keratin-8

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

53.5 kDa

packaging

pkg of 5 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... K8(3856)

General description

The gene K8 (keratin 8) is mapped to human chromosome 12q13.13.[1] It belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with keratin 18.[2][3] The protein has an amino-terminal head, a conserved α-helical central rod domain and a carboxyl-terminal tail domain.[4] The recombinant human keratin-8 is expressed in Escherichia coli and purified by proprietary chromatographic techniques.

Biochem/physiol Actions

K8 (keratin 8) is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas.[5][6][7] Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin polypeptides 8 and 18.[8] Mutations in KRT8 are associated with liver injury as well as liver diseases. It is also an autoantigen of rheumatoid arthritis.[9]

Physical form

5 μg, sterile-filtered and then lyophilized from 30 mM Tris-HCl, pH 8.0, 9.5 M Urea, 2 mM DTT, 2 mM EDTA, and 10 mM methylammonium chloride

Reconstitution

Reconstitute in water to a concentration of 100ug/mL. This solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keratin 8 and 18 mutations are risk factors for developing liver disease of multiple etiologies.
Ku NO
Proceedings of the National Academy of Sciences of the USA, 100, 6063-6068 (2003)
A genome-wide survey over the ChIP-on-chip identified androgen receptor-binding genomic regions identifies a novel prostate cancer susceptibility locus at 12q13.13.
Feng J
Cancer Epidemiology, Biomarkers & Prevention, 20, 2396-2403 (2011)
Cytokeratin 8 immunostaining pattern and E-cadherin expression distinguish lobular from ductal breast carcinoma.
Lehr HA
American Journal of Clinical Pathology, 114, 190-196 (2000)
Keratin 8 is a novel autoantigen of rheumatoid arthritis.
Biochemical and Biophysical Research Communications, 465, 665-669 (2015)
Cancer-associated cleavage of cytokeratin 8/18 heterotypic complexes exposes a neoepitope in human adenocarcinomas.
Ditzel HJ
The Journal of Biological Chemistry, 277, 21712-21722 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service