Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4805

Sigma-Aldrich

Thrombospondin human

recombinant, expressed in baculovirus infected Sf9 cells, ≥98% (SDS-PAGE)

Synonym(s):

THBS, THBS1, TSP, TSP1, Thrombospondin-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥98% (SDS-PAGE)

form

lyophilized powder

mol wt

~180.0 kDa

packaging

pkg of 10 μg
pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TSP1(7057)

General description

The gene TSP1 (thrombospondin 1) is mapped to human chromosome 15q14.[1] It is a matricellular glycoprotein and can be secreted by many cells.[2][3][4] Recombinant human thrombospondin is glycosylated with N-linked sugars and produced using baculovirus vectors in insect cells. Human thrombospondin is purified by proprietary chromatographic techniques.

Application

In rat C6 astroglioma cells, TSP1 (thrombospondin 1)-human has been used in coating the cell culture plates to study the effects of integrins on CNTF (ciliary neurotrophic factor) expression.[5] It has also been used in the preparation of secreted factor cocktail for analysis of the human mesenchymal stem cell secretome.[6]

Biochem/physiol Actions

TSP1 (thrombospondin 1) is believed to play a role in cell migration and proliferation, during embryogenesis and wound repair.[7][8] TSP1 expression is highly regulated by different hormones and cytokines and is developmentally controlled. TSP1 stimulates the growth of vascular smooth muscle cells and human foreskin fibroblasts. A combination of IFN-γ (interferon γ) and TNF-α (tumor necrosis factor α) inhibits TSP1 production in these cells.[9] In endothelial cells, it controls adhesion, migration as well as proliferation. It also exhibits antiangiogenic properties and regulates immune processes.[2][3] TSP1 binds to various cell surface receptors, such as integrins and integrin-associated protein CD47.[7] It also plays a crucial role in inflammatory processes and post-inflammatory tissue dynamics.[4] TSP1 has been used as a potential regulator of tumor growth and metastasis.[3] It is upregulated in rheumatoid synovial tissues and might be associated with rheumatoid arthritis.[10] Variants of this gene might be linked with increased risk of autism.[11]

Physical form

Lyophilized from 20 mM sodium phosphate, pH 6.0, 300
mM NaCl and 0.1% BSA.

Reconstitution

Reconstitute with water at the concentration 0.1 mg/ml. Avoid freeze-thaw cycles.

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of promoter region methylation patterns of MGMT, CDKN2A, GSTP1, and THBS1 genes in intracranial meningioma patients.
Genetic Testing and Molecular Biomarkers, 16, 335-340 (2012)
Thrombospondins as matricellular modulators of cell function.
P Bornstein
The Journal of clinical investigation, 107(8), 929-934 (2001-04-18)
Common and rare variants of the THBS1 gene associated with the risk for autism.
Psychiatric Genetics, 24, 235-240 (2014)
Thrombospondin-1 suppresses wound healing and granulation tissue formation in the skin of transgenic mice.
Streit M
The Embo Journal, 19, 3272-3282 (2000)
Upregulation of Thrombospondin 1 Expression in Synovial Tissues and Plasma of Rheumatoid Arthritis: Role of Transforming Growth Factor-?1 toward Fibroblast-like Synovial Cells.
The Journal of Rheumatology, 42, 943-943 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service