Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4741

Sigma-Aldrich

PLGF-2 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

PGFL, Placenta Growth Factor-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

lyophilized

mol wt

~18.0 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PGF(5228)

General description

PLGF-2 is produced in E. coli as a homodimer containing 2 x 153 amino acids and having a total molecular mass of 36 kDa.

Physical form

Sterile filtered and lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute the protein in sterile 20 mM acetic acid to a concentration of 0.1-1.0 ug/ul. The solution can then be diluted into other aqueous buffer.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service