Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4657

Sigma-Aldrich

Gremlin 2 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

BMP antagonist 2, Cysteine knot superfamily 1, DAN domain family member 3, Gremlin-2, Protein related to DAN and cerberus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~17.2 kDa

packaging

pkg of 20 μg

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GREM2(64388)

General description

Gremlin (also known as Increased in High Glucose Protein 2, IHG-2, Down-regulated in Mos-transformed cells protein, Drm) functions as a bone morphogenetic protein (BMP) antagonist. It acts by binding to, and forming heterodimers with BMP-2, BMP-4, BMP-7, thus preventing them from interacting with their cell surface receptors. Recombinant human Gremlin-2 is a N-terminus His-tagged protein that was expressed from E. coli.

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute to 1 mg/mL with sterilized dH2O. Pipette to dissolve the protein pellet completely.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu Ruiqi et al.
Frontiers in pharmacology, 12, 628671-628671 (2021-11-02)
Peritoneal dialysis (PD) can improve the quality of life of patients with kidney disease and prolong survival. However, peritoneal fibrosis can often occur and lead to PD withdrawal. Therefore, it is imperative to better understand how to inhibit and slow
Ching-Yan Chloé Yeung et al.
Scientific reports, 4, 5183-5183 (2014-06-06)
Tendons are prominent members of the family of fibrous connective tissues (FCTs), which collectively are the most abundant tissues in vertebrates and have crucial roles in transmitting mechanical force and linking organs. Tendon diseases are among the most common arthropathy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service