Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4642

Sigma-Aldrich

VEGF-B human

recombinant, expressed in E. coli

Synonym(s):

VEGFB, Vascular Endothelial Growth Factor-B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥98% (SDS-PAGE)

form

lyophilized

potency

0.5-5.0 μg/mL

mol wt

~19.63 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VEGFB(7423)

General description

Vascular endothelial growth factor B (VEGF-B), a member of the VEGF family,[1] is a potent growth and angiogenic cytokine. VEGF-B is expressed in all tissues except the liver.[2] It forms cell surface-associated disulfide linked homodimers and can form heterodimers with VEGF-A. There are two known isoforms, formed by alternative splicing, which have been designated VEGF-B167 and VEGF-B186. Both forms have identical amino-terminal sequences encoding a "cysteine knot" like structural motif, but differ in their carboxyl-terminal domains. Both VEGF-B isoforms signal only through the VEGFR1 receptor. The gene encoding this protein is localized on human chromosome 11q13 and contains seven exons.[2][3] Recombinant human VEGF-B is a 38kDa disulfide-linked homodimeric protein consisting of two 167 amino acid polypeptide chains.

Biochem/physiol Actions

Vascular endothelial growth factor B (VEGF-B) promotes DNA synthesis in endothelial cells, regulates angiogenesis and vascular permeability, and inhibits apoptosis in certain smooth muscle cells and neurons.[2] It has a role in coronary vessel growth and cardiac hypertrophy, which might provide protection to heart against ischemic damage and heart failure. VEGF-B is also involved in neuroprotection. The protein might have a role in metabolic functions since it is highly expressed in tissues with highly active energy metabolism.[1]

Physical form

Lyophilized without any additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in 0.1% acetic acid to a concentration of 0.1-1.0 mg/mL.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roles of Vascular Endothelial Growth Factor in Amyotrophic Lateral Sclerosis
Ana Catarina
BioMed Research International, 947513-947513 (2014)
Vascular endothelial growth factor-B in physiology and disease.
Bry M et al
Physiological Reviews, 94(3), 779-794 (2014)
Vascular endothelial growth factor and angiogenesis.
Hoeben A et al
Pharmacological Reviews, 56(4), 549-580 (2004)

Questions

  1. Does this protein contain any affinity tag such as HisTag, FLAG tag, StrepTag, or is it a "tag free" version?

    1 answer
    1. The protein does not have a tag according to the supplier.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service