Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4631

Sigma-Aldrich

RELM-alpha from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

Cysteine-rich secreted protein A12-γ, Cysteine-rich secreted protein FIZZ1, Parasite-induced macrophage novel gene 1 protein, RELMα, RELM-a, Resistin-like α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~10.0 kDa

packaging

pkg of 25 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Retna(57262)

General description

RELM-α belongs to a unique family of tissue-specific cytokines termed FIZZ (found in inflammatory zone) and RELM. The three known members of this family; Resistin, RELM-α and RELM-β are 85-94 amino acid secreted proteins sharing a conserved C-terminal domain characterized by 10 cysteine residues with a unique spacing motif of C-X11-C-X8-C-X-C-X3-C-X10-C-X-C-X-C-X9-C-C. RELM-α and Resistin are secreted exclusively by adipocytes while RELM-β is expressed in the epithelium of the colon and small bowel. The physiological role and molecular targets of RELM-α re still unknown. Recombinant mouse RELM-α is a 10.0 kDa monomeric protein containing 88 amino acid residues.

Physical form

Sterile filtered and lyophilized from 10 mM Tris, pH 7.5 plus 25 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1 μg/ul. The solution can then be diluted into other aqueous buffers.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service