Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4593

Sigma-Aldrich

gAcrp30/Adiponectin from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

Acrp30, Adipocyte-specific protein AdipoQ, Adipolean, Apm-1, adipoQ, adiponectin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

assay

≥98% (SDS-PAGE)

form

lyophilized

potency

<1-3 μg/mL

mol wt

~16.6 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

Rat gAcrp30 (amp-1) is a 16.6 kDa protein that corresponds to the C-terminal globular domain of adiponectin (amp-1). gAcrp30/Adipolean is expressed and secreted exclusively by adipose tissue. This protein possesses unique pharmacological properties for regulation of body weight and lipid metabolism. It is a key component in controlling energy homeostasis. Recently, gAcrp30/Adipolean has been shown to inhibit the proliferation of myelomonocytic progenitors. The Recombinant rat gAcrp30/Adipolean is a 16.6 kDa protein consisting of 145 amino acid residues.

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of <0.3 mg/mL. This solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or –20°C for future use.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongyuan Wen et al.
Obesity (Silver Spring, Md.), 28(10), 1932-1940 (2020-08-28)
This study aimed to elucidate the mechanism by which hypertrophic adipocytes regulate insulin signaling in cardiac myocytes. Palmitate was used to induce hypertrophic 3T3-L1 adipocytes. Exosomes were purified from normal control or hypertrophic 3T3-L1 adipocyte-associated conditioned medium. Exosome-exposed neonatal rat

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service