Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4591

Sigma-Aldrich

gAcrp30/Adiponectin human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

Acrp30, Adipocyte-specific protein AdipoQ, Adipolean, Apm-1, adipoQ, adiponectin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥98% (SDS-PAGE)

form

lyophilized

potency

<1-3 μg/mL

mol wt

~16.6 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ADIPOQ(9370)

General description

Acrp30 (adipocyte complement-related protein), or cytokine adiponectin, is a hormone produced and secreted by differentiated adipocytes. It is a 30kDa hydrophilic protein. This protein is composed of an N-terminal collagenous domain and a C-terminal globular trimerization domain. It exists in plasma predominantly as trimeric, hexameric or other high-molecular weight forms. The globular subunit gAcrp30 is however absent in plasma and is biologically active.[1][2] gAcrp30 is the truncated form of full length of Acrp30, and functions as a ligand of AdipoR1.[3]
The Recombinant Human gAcrp30/Adipolean is a 16.6 kDa protein consisting of 145 amino acid residues

Biochem/physiol Actions

In obese mice and humans, adiponectin mRNA expression is reduced, therefore linking this hormone to energy homeostasis. In incubated mouse muscle and cultured cells, gAcrp30 leads to an increase in fatty acid oxidation. This hormone is also linked to anti-atherogenesis, and in apoE (apolipoprotein)-deficient mice, overexpression of gAcrp30 results in significant attenuation of atherosclerosis. Long-term administration of this hormone in mice results in weight loss without reduction in food intake.[1] This hormone is capable of reducing proliferation of MDA-ERα7 cell line. Studies show that ratio of gAcrp30 to leptin and the ERα (estrogen receptor) status of cells are key in determining BRCA (breast cancer) cell growth.[3]

Physical form

Lyophilized from 10 mM Tris, pH 9.2 plus 0.05% Sarcosyl plus 0.75 mM DTT.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 1 mg/mL. This solution can then be diluted into other aqueous buffers.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ACRP30/adiponectin: an adipokine regulating glucose and lipid metabolism.
Berg AH, et al.
Trends in Endocrinology and Metabolism, 13(2), 84-89 (2002)
Metabolic and hormonal interactions between muscle and adipose tissue.
Tomas E et al
The Proceedings of the Nutrition Society, 63(2), 381-385 (2004)
Balance of adiponectin and leptin modulates breast cancer cell growth.
Grossmann ME et al
Cell Research, 18(11), 1154-1156 (2008)
Zhongming Jia et al.
Cell biochemistry and biophysics, 70(2), 1239-1245 (2014-06-08)
Adiponectin, the hormone produced and secreted by adipocytes, has been shown to promote migration of the epithelial cells and angiogenesis in these cells. We sought to determine if adiponectin could induce the cellular migration and growth factor expression in breast

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service