Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4581

Sigma-Aldrich

BMP-10 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

BMP 10, bone morphogenetic protein 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~32 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BMP10(27302)

General description

BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. As implied by their name, BMPs promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. BMP-10 is essential for the proper development of the heart, but is not expressed until after cardiac patterning or looping are completed. Recombinant human BMP-10 is a 32 kDa disulfide-linked homodimer containing two 142 amino acid polypeptide chains.
The bone morphogenetic protein 10 (BMP10) gene spans ~7.8 kb on human chromosome 2p13.3.[1] It is highly expressed in the trabeculae of the embryonic heart, but it is expressed at low levels in the liver and the lungs.[1] BMP-10 belongs to the bone morphogenetic protein (BMP) family.[1]

Biochem/physiol Actions

Bone morphogenetic protein 10 (BMP10) acts as a potent tumor suppressor in prostate, gastric and breast cancer.[1][2] It is implicated in trabeculation of the embryonic heart. BMP10 retards the growth of prostate cancer cells via, apoptosis induced by X- inhibitor of apoptosis protein (XIAP) and extracellular signal–regulated kinase (ERK)1/2 dependent signaling and SMAD-independent signaling. In addition, it also has an ability to inhibit prostate cancer cell migration and invasiveness.[1] Elevated expression of the gene inhibits growth, adhesion and migration of bladder cancer cells in vitro. Thus, this protein can be considered as a potent therapeutic target for bladder cancer.[1]

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute to a concentration of 0.1-1.0 mg/mL in water containing BSA (50 mg BSA per 1 mg of protein). This solution can then be diluted into other aqueous buffers

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BMP10 inhibited the growth and migration of gastric cancer cells.
Lei H, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 37(3), 3025-3031 (2016)
Jinfu Zhuang et al.
Biological chemistry, 401(2), 285-296 (2019-06-13)
The gene encoding transmembrane protein 100 (TMEM100) was first discovered to be transcribed by the murine genome. It has been recently proven that TMEM100 contributes to hepatocellular carcinoma and non-small-cell lung carcinoma (NSCLC). This study investigates the impact of TMEM100
Lijun Sun et al.
Journal of cellular biochemistry, 115(11), 1868-1876 (2014-06-07)
Heart disease is among the leading causes of death worldwide, and the limited proliferation of mammalian cardiomyocytes prevents heart regeneration in response to injury. Bone morphogenetic protein-10 (BMP10) exerts multiple roles in various developmental events; however, the effect of BMP10
Ning Zhang et al.
Anticancer research, 33(5), 1917-1925 (2013-05-07)
Bone morphogenetic protein-10 (BMP10), a novel member of the bone morphogenetic protein (BMP) family, has been indicated as a possible tumour suppressor in prostate and breast cancer. However, its role in urothelial tumours remains unknown. In the present study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service