Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4548

Sigma-Aldrich

IL-20 human

recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)

Synonym(s):

D ZCYTO10, Four α helix cytokine, IL-20, IL10, MGC96907, Zcyto10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

human

recombinant

expressed in E. coli

assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

mol wt

~17.6 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL20(50604)

General description

Interleukin-20 has a 28% amino acid identity with IL-10 and induces kerotinocyte proliferation. Recombinant human IL-20 is a 17.6 kDa protein consisting of 153 amino acids residues.

Physical form

Lyophilized from 10 mM Sodium Citrate, pH 3.0 plus 100 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1 to 1.0 μg/μL. It is recommended that further dilutions be made into buffer containing carrier protein or medium containing serum.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service