Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4507

Sigma-Aldrich

FABP7 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

B-FABP, Brain lipid-binding protein, Brain-type fatty acid-binding protein, Fatty acid-binding protein 7, Fatty acid-binding protein brain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

~16 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FABP7(2173)

General description

FABPs have been reported to be involved in the uptake and metabolism of fatty acids, maintenance of cellular membrane fatty acids levels, intracellular trafficking, modulation of specific enzymes of lipid metabolic pathways, as well as in the modulation of cell growth and differentiation.

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pavel Gromov et al.
PloS one, 9(11), e112024-e112024 (2014-11-13)
Apocrine carcinoma of the breast is a distinctive malignancy with unique morphological and molecular features, generally characterized by being negative for estrogen and progesterone receptors, and thus not electable for endocrine therapy. Despite the fact that they are morphologically distinct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service