Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4497

Sigma-Aldrich

Eotaxin-2/CCL24 human

recombinant, expressed in E. coli, ≥97% (SDS-PAGE)

Synonym(s):

C-C motif chemokine 24, CK-β-6, Eosinophil chemotactic protein 2, Eotaxin-2, Myeloid progenitor inhibitory factor 2, Small-inducible cytokine A24

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥97% (SDS-PAGE)

form

lyophilized

potency

10 ng/mL

mol wt

~8.8 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL24(6369)

General description

Eotaxin-2 also known as MPIF-2 is a CC chemokine characterized by its high chemotactic selectivity for eosinophils. Human eotaxin-2 is 8.8 kDa protein containing 78 amino acid residues.

Physical form

The protein was lyophilized from a concentrated (1 mg/ml) sterile solution containing 20 mM PBS pH-7.4 and 0.15 M sodium chloride.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1 μg/μL. It is recommended that further dilutions be made into buffer containing carrier protein or medium containing serum.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service