Stem Cell Factor (SCF) is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis. SCF stimulates the proliferation of myeloid, erythroid, and lymphoid progenitors in bone marrow cultures. Recombinant rat SCF is an 18.4 kDa protein containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF.
Physical form
Lyophilized from 20 mM Tris, pH 7.5.
Reconstitution
Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.
Proceedings of the National Academy of Sciences of the United States of America, 112(31), 9638-9643 (2015-07-22)
Determining the developmental pathway leading to erythrocytes and being able to isolate their progenitors are crucial to understanding and treating disorders of red cell imbalance such as anemia, myelodysplastic syndrome, and polycythemia vera. Here we show that the human erythrocyte
Asian-Australasian journal of animal sciences, 28(11), 1565-1572 (2015-11-19)
Porcine embryonic stem cells (pESCs) have become an advantageous experimental tool for developing therapeutic applications and producing transgenic animals. However, despite numerous reports of putative pESC lines, deriving validated pESC lines from embryos produced in vitro remains difficult. Here, we
The Journal of clinical investigation, 125(8), 3103-3116 (2015-07-21)
Severe congenital neutropenia (SCN) is often associated with inherited heterozygous point mutations in ELANE, which encodes neutrophil elastase (NE). However, a lack of appropriate models to recapitulate SCN has substantially hampered the understanding of the genetic etiology and pathobiology of
Hematopoietic stem cells (HSC) differentiate into megakaryocytes (MK), whose function is to release platelets. Attempts to improve in vitro platelet production have been hampered by the low amplification of MK. Providing HSC with an optimal three-dimensional (3D) architecture may favor
International journal of radiation biology, 90(7), 594-604 (2014-03-07)
Amifostine is a highly efficacious cytoprotectant when administered in vivo at high doses. However, at elevated doses, drug toxicity manifests for general, non-clinical radioprotective purposes. Various strategies have been developed to avoid toxic side-effects: The simplest is reducing the dose.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.