Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4260

Sigma-Aldrich

MIP-5/CCL15 human

recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)

Synonym(s):

CCL15, Chemokine CC-2, MIP5, Macrophage inflammatory protein 5, Small inducible cytokine A15 precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

mol wt

~10.1 kDa

packaging

pkg of 25 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL15(6359)

General description

Human macrophage inflammatory protein-5 (MIP-5) also known as Lkn-1 has been shown to be chemotactic for T-lymphocytes but not for monocytes or granulocytes. Recombinant human MIP-5β is a 10.1 kDa protein containing 92 amino acid residues.

Physical form

Lyophilized from a concentrated (1 mg/ml) solution containing 20 mM PBS pH7.4 and 100 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service