Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4257

Sigma-Aldrich

MIP-4 (PARC) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CC chemokine PARC, CCL18, MIP-4, Macrophage inflammatory protein 4, Pulmonary and activation-regulated chemokine, Small inducible cytokine A18

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~7.8 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL18(6362)

General description

Human microphage inflammatory protein-4 (MIP-4) also known as PARC, has been shown to be chemotactic for T lymphocytes but not for monocytes or granulocytes. Human MIP-4 is an 7.8 kDa protein containing 68 amino acid residues.

Physical form

Sterile filtered and then lyophilized without any additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 1 mg/mL. The solution can be diluted into other buffered solutions.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service