Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4178E

Sigma-Aldrich

IL-17E from rat

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

IL17E, IL25, InterleukinE

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

~35.5 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

Rat IL-17E (IL-17E), is a proinflammatory cytokine member of a six-species family of proteins (IL-17A-17F). Rat IL-17E protein is a homodimer consisting of two 145 amino acids peptides. Recombinant rat IL-17E produced in E. coli is a non-glycosylated disulfide-joined homodimer having a molecular mass of 35.5 kDa.

Physical form

Lyopohilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute the lyophilized product with sterile 10 mM HCl at a concentration of 0.1 mg′s/ml. Further dilutions into other buffers can be made.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service