Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4107

Sigma-Aldrich

GRO-gamma human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CXCL3, GRO-γ, GRO-γ(1-73), Growth-regulated protein γ, MIP2-β, Macrophage inflammatory protein 2-β

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

biological source

human

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

10-100 ng/mL

mol wt

~7.9 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CXCL3(2921)

General description

Human GRO-γ is a member of the C-X-C family of chemokines and promotes neutrophil and basophil chemotaxis and degranulation. Recombinant human GRO-γ is a 7.9 kDa protein containing 73 amino acid residues.

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service