Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4058

Sigma-Aldrich

Fgf-9 from rat

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

FGF9, GAF, Glia-activating factor, HBGF-9, fibroblast growth factor 9

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~23.3 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Fgf9(25444)

General description

FGF-9 (also known as Glial Activating Factor) is a heparin binding growth factor that stimulates the proliferation and activation of glial cells and other cells that express FGF receptors. The rat FGF-9 is a 23.3 kDa protein containing 205 amino acid residues.

Physical form

Lyophilized from 10 mM Tris, pH 7.8 plus Ammmonium Sulfate.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in 10 mM Tris (pH 8.0) to a concentration of 0.1-1.0 mg/mL. This solution can be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service