SRP4028
Eotaxin human
recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)
Synonym(s):
CCL11, Eosinophil chemotactic protein chemokine (C-C motif) ligand 11, MGC22554, SCYA11, Small inducible cytokine A11
About This Item
Recommended Products
biological source
human
recombinant
expressed in E. coli
assay
≥97% (HPLC)
≥97% (SDS-PAGE)
form
lyophilized
potency
10 ng/mL
mol wt
~8.3 kDa
packaging
pkg of 20 μg
storage condition
avoid repeated freeze/thaw cycles
impurities
endotoxin, tested
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CCL11(6356)
General description
Biochem/physiol Actions
Physical form
Reconstitution
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service