Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4025

Sigma-Aldrich

ENA-78 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CXCL5, ENA-78, Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide, Small inducible cytokine B5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

50 ng/mL

mol wt

~8 kDa

packaging

pkg of 20 μg

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CXCL5(6374)

General description

ENA-78 (Epithelial Neutrophil Activating Peptide-78) is a recently discovered chemoattractant and activator for neutrophils, belonging to the IL-8 subgroup of the C-X-C family of chemokines. The human ENA-78 is an 8.0 kDa protein containing 74 amino acid residues.

Physical form

Lyophilized with no additives. The protein may appear as a haze or film which is difficult to see at the bottom of the vial.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1 μg/μL. It is recommended that further dilutions be made into buffer containing carrier protein or medium containing serum

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiyuki Mitsuyama et al.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.], 15(3), 271-280 (2015-03-31)
Characteristics of type 2 autoimmune pancreatitis (AIP) is granulocyte epithelial lesions, called idiopathic duct-centric pancreatitis (IDCP). To clarify pathogenesis of IDCP, we investigated mechanism of neutrophil infiltration in type 1 AIP, called lymphoplasmacytic sclerosing pancreatitis (LPSP) and IDCP. This study
Debjani Guha et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 35(5), 373-384 (2014-12-24)
Human immunodeficiency virus-1 (HIV-1)-infected monocytes/macrophages and microglia release increased levels of proinflammatory cytokines and chemokines, including ELR+ (containing glutamic acid-leucine-arginine motif) chemokines. To investigate the role of HIV-1 infection on chemokine regulation, monocyte-derived macrophages (MDMs) from normal donors were infected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service