Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3319

Sigma-Aldrich

EGF-L7 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE or HPLC), ≥98% (HPLC)

Synonym(s):

MEGF7, Multiple EGF-like domains protein 7

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$347.00

$347.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
$347.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$347.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE or HPLC)

form

lyophilized

mol wt

28.0 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EGFL7(51162)

General description

EGF-L7 (Epidermal Growth Factor Like Protein 7, Multiple EGF Like Domains Protein 7, VE Statin) is a multi-domain protein containing two EGF like domains and one EMI domain. It is expressed almost exclusively in endothelial cells and functions to promote normal development of the vascular system, particularly tubulogenesis. EGF-L7 is capable of antagonistic binding to Notch receptors resulting in the inhibition of Notch signaling in HUVEC and neural stem cells. In research models inducing hypoxia and subsequent reoxygenation, (H/R), EGF-L7 can inhibit ICAM-1 expression and enhance the inhibition of NF-kappaB activation. Additionally, EGF-L7 can chemoattract endothelial cells and bind to the extracellular matrix. The overexpression of EGF-L7 is observed in various cancers, and is generally correlated with increased metastasis and a poor prognosis. Recombinant human EGF-L7 is a 28 kDa protein containing 251 amino acid residues.

Biochem/physiol Actions

EGF-L7 (Epidermal Growth Factor Like Protein 7, Multiple EGF Like Domains Protein 7, VE Statin) is a multi-domain protein containing two EGF like domains and one EMI domain. Recombinant human EGF-L7 is a 28 kDa protein containing 251 amino acid residues.

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luisa Campagnolo et al.
The American journal of pathology, 167(1), 275-284 (2005-06-24)
The endothelium of the adult vasculature is normally quiescent, with the exception of the vasculature of the female reproductive system. However, in response to appropriate stimuli (ie, wound healing, atherosclerosis, tumor growth and metastasis, arthritis) the vasculature becomes activated and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service