Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3307

Sigma-Aldrich

FGF-8 human

Animal-component free, recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)

Synonym(s):

AIGF, FGF-8b, HBGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

human

recombinant

expressed in E. coli

assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

potency

2.0-4.0 ng/mL

mol wt

22.5 kDa

packaging

pkg of 25 μg

impurities

endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF8(2253)

General description

FGF-8 (FGF-8b) is a heparin binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF8A, FGF-8B, FGF-8E and FGF-8F. The human and murine FGF8 A and B are identical unlike human and mouse FGF8 E and F are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors. Recombinant human FGF-8 (FGF-8b) is a 22.5 kDa protein consisting of 194 amino acid residues.

Biochem/physiol Actions

FGF-8 (FGF-8b) is a heparin binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. . Recombinant human FGF-8 (FGF-8b) is a 22.5 kDa protein consisting of 194 amino acid residues.

Physical form

Lyophilized from 10 mM Sodium Phosphate pH, 8.0 + 75 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fibroblast growth factors.
Ornitz, D.M. and Itoh, N.
Genome Biology, 2, 3005-3005 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service