Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3232

Sigma-Aldrich

RELM-beta from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Cysteine-rich secreted protein FIZZ2, Resistin-like beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

18.0 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.2 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... RETNLB(57263)

General description

Resistin-like molecule-β (RELM-β), also known as found in inflammatory zone-2 (FIZZ-2), is a member of RELM/FIZZ family of cysteine-rich secretory proteins. Recombinant murine RELM β is an 18.0kDa, consisting of two 83 amino acid residue chains. RELM-β contains highly conserved C-terminal domain and the molecular structure is highly homologous to that of adipose-derived cytokine resistin.[1] It is a disulfide-linked homodimeric protein synthesized and secreted by goblet cells in intestine.[2] RELM-β is expressed in the lumen of the bowel and epithelial cells of the intestine.[1][3]
Recombinant Murine RELM beta is an 18.0kDa, consisting of two 83 amino acid residue chains.

Biochem/physiol Actions

Increased expression of resistin-like molecule-β (RELM-β) in asthmatic airways is associated with pulmonary inflammation and airways remodeling in the mouse. It increases deposition of matrix proteins via enhancing fibroblast proliferation and differentiation.[1] RELM-β is implicated in interleukin 13 (IL-13) induced antiparasitic activity. RELM-β plays a vital role in the regulation of colonic epithelial cell barrier function and gastrointestinal innate immunity. Decreased expression of RELM-β leads to colonic inflammation.[4]

Sequence

MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA

Physical form

Lyophilized from 10 mM Acetic Acid.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resistin-like molecule-? (RELM-?) targets airways fibroblasts to effect remodelling in asthma: from mouse to man.
Fang CL
Clinical and Experimental Allergy, 45(5), 940-952 (2015)
Intestinal goblet cells and mucins in health and disease: recent insights and progress.
Kim YS and Ho SB.
Current Gastroenterology Reports, 12(5), 319-330 (2010)
Correlation between serum resistin level and adiposity in obese individuals.
Azuma K
Obesity Research, 11(8), 997-1001 (2003)
Rijala, M., et al.
The Journal of Clinical Investigation, 3, 225-230 (2003)
Resistin-like molecule beta regulates innate colonic function: barrier integrity and inflammation susceptibility.
Hogan SP
The Journal of Allergy and Clinical Immunology, 118(1), 257-268 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service