Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3193

Sigma-Aldrich

Cardiotrophin-1 from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CT-1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$382.00

$382.00


Usually ships in 2 weeks.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
10 μG
$382.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$382.00


Usually ships in 2 weeks.

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤1.0 ng/mL ED50

mol wt

21.3 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

off-white to yellow

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... CTF1(13019)

General description

Cardiotrophin-1 is from interleukin (IL)-6 family of cytokines. It is mainly present in adipose tissues, osteoclasts and cardiomyocytes.[1][2][3] Cardiotrophin-1 works through gp130 (glycoprotein) and the LIFR (LIF receptor).[2]

Biochem/physiol Actions

In adipocytes, cardiotrophin-1 helps in activating signaling pathways associated with metabolism. It controls energy homeostasis, and glucose and lipid metabolism. High levels of cardiotrophin-1 are reported in obesity and metabolic syndrome.[1] Absence of cardiotrophin-1 in mice causes mature-onset obesity, insulin resistance and hypercholesterolemia.[4] In osteoclasts, it enhances osteoblast activity as well as mineralization.[2] It also participates in cardiomyocyte survival, hypertrophy, cardiac fibroblast proliferation and collagen type I synthesis.[5] Cardiotrophin-1 is associated with arterial fibrosis and stiffness.[3]

Sequence

SQREGSLEDH QTDSSISFLP HLEAKIRQTH NLARLLTKYA EQLLEEYVQQ QGEPFGLPGF SPPRLPLAGL SGPAPSHAGL PVSERLRQDA AALSVLPALL DAVRRRQAEL NPRAPRLLRS LEDAARQVRA LGAAVETVLA ALGAAARGPG PEPVTVATLF TANSTAGIFS AKVLGFHVCG LYGEWVSRTE GDLGQLVPGG VA

Physical form

Lyophilized from 0.5mg/ml in 5mM Tris, pH 8.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1- 1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A role for cardiotrophin-1 in myocardial remodeling induced by aldosterone.
Lopez-Andres N, et al.
American Journal of Physiology. Heart and Circulatory Physiology, 301, H2372-H2372 (2011)
Cardiotrophin-1 is a key regulator of glucose and lipid metabolism.
Moreno-Aliaga MJ et al.
Cell Metabolism, 14, 242-242 (2011)
Cardiotrophin-1 is a key regulator of glucose and lipid metabolism.
Moreno-Aliaga MJ, et al.
Cell Metabolism, 14 (2011)
Cardiotrophin-1 stimulates lipolysis through the regulation of main adipose tissue lipases.
Lopez-Yoldi M et al.
Journal of Lipid Research, 55, 2634-2634 (2014)
Absence of cardiotrophin 1 is associated with decreased age-dependent arterial stiffness and increased longevity in mice.
Lopez-Andres N, et al.
Hypertension, 61, 120-120 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service