Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3176

Sigma-Aldrich

TLR-3 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

CD283 antigen, Toll-like receptor 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

≤30 μg/mL ED50

mol wt

77.9 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TLR3(7098)

General description

TLR-3 is a single-pass type I receptor that binds to and signals the presence of microbial pathogens and double stranded RNA (dsRNA) viruses. Signaling through TLR-3 can promote the NF-kB pathway to initiate innate and adaptive immune responses to bacterial and viral infections, as well as the p53 pathway to trigger apoptosis in cells infected with dsRNA viruses. TLR-3 belongs to a family of structurally-related toll-like receptors (TLRs) containing an N-terminal domain rich in leucine repeats and a C-terminal intracellular Toll/interleukin (IL)-1 (TIL) domain. TLR-3 is expressed primarily in dendritic cells of the placenta and pancreas where it can reside on both sides of the plasma membrane and in the endosomal compartment of the cells. Recombinant human TLR-3 is 77.0 kDa glycoprotein containing 681 residues which comprise the TLR-3 extracellular domain.

Biochem/physiol Actions

TLR-3 is a single-pass type I receptor that binds to and signals the presence of microbial pathogens and double stranded RNA (dsRNA) viruses. Recombinant human TLR-3 is 77.0 kDa glycoprotein containing 681 residues which comprise the TLR-3 extracellular domain.

Sequence

STTKCTVSHE VADCSHLKLT QVPDDLPTNI TVLNLTHNQL RRLPAANFTR YSQLTSLDVG FNTISKLEPE LCQKLPMLKV LNLQHNELSQ LSDKTFAFCT NLTELHLMSN SIQKIKNNPF VKQKNLITLD LSHNGLSSTK LGTQVQLENL QELLLSNNKI QALKSEELDI FANSSLKKLE LSSNQIKEFS PGCFHAIGRL FGLFLNNVQL GPSLTEKLCL ELANTSIRNL SLSNSQLSTT SNTTFLGLKW TNLTMLDLSY NNLNVVGNDS FAWLPQLEYF FLEYNNIQHL FSHSLHGLFN VRYLNLKRSF TKQSISLASL PKIDDFSFQW LKCLEHLNME DNDIPGIKSN MFTGLINLKY LSLSNSFTSL RTLTNETFVS LAHSPLHILN LTKNKISKIE SDAFSWLGHL EVLDLGLNEI GQELTGQEWR GLENIFEIYL SYNKYLQLTR NSFALVPSLQ RLMLRRVALK NVDSSPSPFQ PLRNLTILDL SNNNIANIND DMLEGLEKLE ILDLQHNNLA RLWKHANPGG PIYFLKGLSH LHILNLESNG FDEIPVEVFK DLFELKIIDL GLNNLNTLPA SVFNNQVSLK SLNLQKNLIT SVEKKVFGPA FRNLTELDMR FNPFDCTCES IAWFVNWINE THTNIPELSS HYLCNTPPHY HGFPVRLFDT SSCKDSAPFE L

Physical form

Lyophilized from 20 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Schröder et al.
Trends in immunology, 26(9), 462-468 (2005-07-20)
Toll-like receptor 3 (TLR3), which recognizes double-stranded (ds)RNA, was the first identified antiviral TLR and, because dsRNA is a universal viral molecular pattern, TLR3 has been assumed to have a central role in the host response to viruses. However, this
Manabu Taura et al.
Molecular and cellular biology, 28(21), 6557-6567 (2008-09-10)
Toll-like receptors (TLRs) are important sensors of microbial pathogens and mediators of innate immune responses. Although the signal transduction of TLRs is well elucidated, their basal regulation is largely unexplored. Here we show that the tumor suppressor p53 positively regulates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service