Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3170

Sigma-Aldrich

TGF-beta 2 human

recombinant, expressed in insect cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

BSC-1, Cetermin, Glioblastoma-derived T cell suppressor factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

recombinant

expressed in insect cells

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

25.0 kDa

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TGFB2(7042)

General description

The three mammalian isoforms of TGF-b, TGF-b1, b2, b3, signal through the same receptor and elicit similar biological responses. They are multifunctional cytokines that regulate cell proliferation, growth, differentiation and motility as well as synthesis and deposition of the extracellular matrix. They are involved in various physiological processes including embryogenesis, tissue remodeling and would healing. They are secreted predominantly as latent complexes which are stored at the cell surface and in the extracellular matrix. The release of biologically active TGF-b isoform from a latent complex involves proteolytic processing of the complex and /or induction of conformational changes by proteins such as thrombospondin-1. TGF-b2 has been shown to exert suppressive effects on IL-2 dependent T-cell growth, and may also have an autocrine function in enhancing tumor growth by suppressing immuno-surveillance of tumor development. Recombinant human TGF-b2 is a 25.0 kDa protein composed of two identical 112 amino acid polypeptide chains linked by a single disulfide bond.

Biochem/physiol Actions

The three mammalian isoforms of TGF-b, TGF-b1, b2, b3, signal through the same receptor and elicit similar biological responses. Recombinant human TGF-b2 is a 25.0 kDa protein composed of two identical 112 amino acid polypeptide chains linked by a single disulfide bond.

Sequence

ALDAAYCFRN VQDNCCLRPL YIDFKRDLGW KWIHEPKGYN ANFCAGACPY LWSSDTQHSR VLSLYNTINP EASASPCCVS QDLEPLTILY YIGKTPKIEQ LSNMIVKSCK CS

Physical form

Lyophilized from Sodium Citrate, pH 3.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1- 1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen F Chambers et al.
PloS one, 6(4), e18796-e18796 (2011-05-03)
Cell shape and tissue architecture are controlled by changes to junctional proteins and the cytoskeleton. How tissues control the dynamics of adhesion and cytoskeletal tension is unclear. We have studied epithelial tissue architecture using 3D culture models and found that
Flanders, K.C., et al.
Cytokine Reference, 719-746 (2001)
Selen Catania Muratoglu et al.
PloS one, 6(12), e28846-e28846 (2011-12-17)
Vascular remodeling in response to alterations in blood flow has been shown to modulate the formation of neo-intima. This process results from a proliferative response of vascular smooth muscle cells and is influenced by macrophages, which potentiate the development of
Uma S Kamaraj et al.
Cell systems, 11(5), 509-522 (2020-10-11)
The need to derive and culture diverse cell or tissue types in vitro has prompted investigations on how changes in culture conditions affect cell states. However, the identification of the optimal conditions (e.g., signaling molecules and growth factors) required to maintain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service