Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3116

Sigma-Aldrich

MIP-3 (CCL23) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CCL23, Ck-b8, MPIF-1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$365.00

$365.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
$365.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$365.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

11.3 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL23(6368)

General description

MIP-3 is a CC chemokine that signals through the CCR1 receptor. MIP-3 chemoattracts monocytes, resting T-lymphocytes and neutrophils, but does not chemoattract activated lymphocytes. Additionally, MIP-3 has been shown to inhibit colony formation of bone marrow myeloid immature progenitors. Recombinant human MIP-3 is an 11.3 kDa protein containing 99 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.

Biochem/physiol Actions

MIP-3 is a CC chemokine that signals through the CCR1 receptor. Recombinant human MIP-3 is an 11.3 kDa protein containing 99 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.

Sequence

RVTKDAETEF MMSKLPLENP VLLDRFHATS ADCCISYTPR SIPCSLLESY FETNSECSKP GVIFLTKKGR RFCANPSDKQ VQVCMRMLKL DTRIKTRKN

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Hromas et al.
Blood, 89(9), 3315-3322 (1997-05-01)
Chemokines are a family of related proteins that regulate leukocyte infiltration into inflamed tissue. Some chemokines such as MIP-1 alpha also inhibit hematopoietic progenitor cell proliferation. Recently, three chemokines, MIP-1 alpha, MIP-1 beta, and RANTES, have been found to significantly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service