Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3035

Sigma-Aldrich

Exodus-2 (CCL21) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

6Ckine, CCL21, SLC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

10.0-100.0 ng/mL

mol wt

12.2 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL21(6366)

General description

Exodus-2 or C-C motif chemokine ligand 21 (CCL21) is a CC chemokine that signals through the C-C chemokine receptor-7 (CCR7). It is expressed in lymph nodes of certain endothelial cells, and in the spleen and appendix.[1][2] CCL21 contains six cysteine residues, including the four conserved cysteines present in CC chemokines. The gene encoding this protein is localized on human chromosome 9p13.[2] Recombinant human Exodus-2 is a 12.2kDa protein containing 111 amino acid residues.

Biochem/physiol Actions

Exodus-2 aids in the migration of lymphocytes and dendritic cells to lymph nodes. This is observed in ulcerative colitis. It also chemoattracts T and B lymphocytes and inhibits hematopoiesis.[1] The protein may have a role in the metastasis of pancreatic cancer stem cells.[3] CCL21 has been associated with lymph node metastasis in gastric carcinoma.[4]

Sequence

SDGGAQDCCL KYSQRKIPAK VVRSYRKQEP SLGCSIPAIL FLPRKRSQAE LCADPKELWV QQLMQHLDKT PSPQKPAQGC RKDRGASKTG KKGKGSKGCR KTERSQTPKG P

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of secondary lymphoid tissue
chemokine suppression on experimental
ulcerative colitis in mice
H. Zhang
Genetics and molecular research : GMR, 13(2) (2014)
[CCL21 promotes the metastasis of human pancreatic cancer Panc-1 cells via epithelial- mesenchymal transition].
Liu Q
Xi bao yu fen zi mian yi xue za zhi = Chinese journal of cellular and molecular immunology, 31(1), 6-9 (2015)
Expression of Chemokine Receptor CCR7 Is Associated with Lymph Node Metastasis of Gastric Carcinoma
Kohjiro Mashino
Cancer Research, 62(10) (2002)
J A Hedrick et al.
Journal of immunology (Baltimore, Md. : 1950), 159(4), 1589-1593 (1997-08-15)
The chemokines are emerging as a diverse and expanding family of cytokines. In this study, we report the identification of a novel beta (CC) chemokine that exhibits an unusual pattern of six conserved cysteines and was thus designated 6Ckine. The
Expression of chemokine receptor CCR7 is associated with lymph node metastasis of gastric carcinoma.
Mashino K, et al.
Cancer Research, 62(10), 2937-2941 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service