SRP2162
PCAF (p300/CBP-Associated Factor) human
recombinant, expressed in insect cells, ≥70% (SDS-PAGE)
Synonym(s):
CAF, P, P/CAF, PCAF
About This Item
biological source
human
recombinant
expressed in insect cells
assay
≥70% (SDS-PAGE)
form
frozen liquid
mol wt
~94.8 kDa
packaging
pkg of 5 μg
concentration
500 μg/mL
color
clear colorless
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... KAT2B(8850)
General description
Biochem/physiol Actions
Physical form
Preparation Note
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service