Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2136

Sigma-Aldrich

USF1, GST tagged human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

FCHL, FCHL1, HYPLIP1, MLTF, MLTFI, UEF, bHLHb11

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~60 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

1000 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... USF1(7391)

Biochem/physiol Actions

Human Upstream Stimulatory Factor 1 (USF1) regulates the transcription of many genes involved in lipid and glucose homeostasis and co-localizes with familial combined hyperlipidemia (FCHL) and type 2 diabetes genes on chromosome 1q22-23. The USF1 protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. It is a basic helix-loop-helix leucine zipper (b-HLH-ZIP) protein that shares a common DNA-binding specificity with the c-Myc oncoproteins.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Du et al.
The EMBO journal, 12(2), 501-511 (1993-02-01)
Earlier in vitro studies identified USF as a cellular factor which activates the adenovirus major late (Ad-ML) promoter by binding to an E-box motif located at position -60 with respect to the cap site. Purified USF contains 44 and 43
Wendy Putt et al.
Human molecular genetics, 13(15), 1587-1597 (2004-06-04)
Upstream stimulatory factor 1 (USF 1), is a transcription factor controlling expression of several genes involved in lipid and glucose homeostasis and co-localizes with familial combined hyperlipidemia (FCHL) and type 2 diabetes on chromosome 1q22-23. We sequenced USF1 in 24
Päivi Pajukanta et al.
Nature genetics, 36(4), 371-376 (2004-03-03)
Familial combined hyperlipidemia (FCHL), characterized by elevated levels of serum total cholesterol, triglycerides or both, is observed in about 20% of individuals with premature coronary heart disease. We previously identified a locus linked to FCHL on 1q21-q23 in Finnish families

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service