Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2085

Sigma-Aldrich

BCL-10 human

recombinant, expressed in insect cells, ≥80% (SDS-PAGE)

Synonym(s):

CARMEN, CIPER, CLAP, c-E10, mE10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

biological source

human

recombinant

expressed in insect cells

assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~28 kDa

packaging

pkg of 6 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

250 μg/mL

color

clear
colorless

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BCL10(8915)

Biochem/physiol Actions

Bcl-10 is an intracellular signaling protein identified from the t(1;14)(p22;q32) breakpoint in MALT lymphomas. The human Bcl10 gene encodes a protein of 233 amino acids containing an N-terminal caspase recruitment domain (CARD) that mediates self-oligomerization and the C-terminal region of Bcl-10. Bcl 10 is expressed ubiquitously with high expression levels in lymphoid tissues and in the developing central nerve system. Mutations have been found in cases of follicular lymphoma and diffuse large B cell lymphoma. Bcl-10 is a positive regulator of antigen-receptor induced NF-kB activation and Bcl-10-initiated activation of NF-kB can be inhibited by cotransfection of dominant-negative mutants of TRAF2, NIK, IKKa or IkBa.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Srinivasula et al.
The Journal of biological chemistry, 274(25), 17946-17954 (1999-06-11)
Molecules that regulate NF-kappaB activation play critical roles in apoptosis and inflammation. We describe the cloning of the cellular homolog of the equine herpesvirus-2 protein E10 and show that both proteins regulate apoptosis and NF-kappaB activation. These proteins were found
T G Willis et al.
Cell, 96(1), 35-45 (1999-02-16)
MALT B cell lymphomas with t(1;14)(p22;q32) showed a recurrent breakpoint upstream of the promoter of a novel gene, Bcl10. Bcl10 is a cellular homolog of the equine herpesvirus-2 E10 gene: both contain an amino-terminal caspase recruitment domain (CARD) homologous to
Q Zhang et al.
Nature genetics, 22(1), 63-68 (1999-05-13)
Mucosa-associated lymphoid tissue (MALT) lymphomas most frequently involve the gastrointestinal tract and are the most common subset of extranodal non-Hodgkin lymphoma (NHL). Here we describe overexpression of BCL10, a novel apoptotic signalling gene that encodes an amino-terminal caspase recruitment domain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service