Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2074

Sigma-Aldrich

Vitamin D receptor (118-427), His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

NR1I1, vitamin D (1,25- dihydroxyvitamin D3) receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~37 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

550 μg/mL

technique(s)

deacetylase assay: suitable

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... VDR(7421)

Biochem/physiol Actions

The vitamin D endocrine system is critical for the proper development and maintenance of mineral ion homeostasis and skeletal integrity. Beyond these classical roles, recent evidence suggests that the bioactive metabolite of vitamin D, 1,25-dihydroxyvitamin D3, functions in diverse physiological processes, such as hair follicle cycling, blood pressure regulation, and mammary gland development. The biological effects of 1,25-(OH)2D3 are mediated through the vitamin D receptor (VDR), a member of the nuclear receptor superfamily of ligand-activated transcription factors. The cellular effects of VDR signaling include growth arrest, differentiation and/or induction of apoptosis. VDR heterodimerizes with RXR and the liganded VDR-RXR heterodimer binds with high affinity to vitamin D response elements (VDREs) in the promoters of target genes. In addition, several nuclear receptor coactivators (SRC-1, DRIP) have been shown to interact with VDR and potentiate its transcriptional activity. In addition to treating disorders of mineral metabolism and diseases of the skeleton, such as rickets, osteoporosis, and renal osteodystrophy, VDR and 1,25-(OH)2D3 have significant therapeutic potential for pathologies such as cancer, autoimmune syndromes, and psoriasis.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amelia L M Sutton et al.
Molecular endocrinology (Baltimore, Md.), 17(5), 777-791 (2003-03-15)
The vitamin D endocrine system is critical for the proper development and maintenance of mineral ion homeostasis and skeletal integrity. Beyond these classical roles, recent evidence suggests that the bioactive metabolite of vitamin D, 1,25-dihydroxyvitamin D3, functions in diverse physiological
Brown, A.J., et al.
The American Journal of Physiology, 277-277 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service