Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2062

Sigma-Aldrich

STAT3 human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

APRF, FLJ20882, HIES, MGC16063

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in E. coli

assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~90 kDa

storage condition

avoid repeated freeze/thaw cycles

concentration

400 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... STAT3(6774)

Biochem/physiol Actions

Signal transducer and activator of transcription (STAT) proteins are a family of latent cytoplasmic transcription factors involved in cytokine, hormone, and growth factor signal transduction. Seven members of the STAT family of transcription factors have been identified in mammalian cells: STAT1, STAT2, STAT3, STAT4, STAT5a, STAT5b, and STAT6. STAT proteins mediate broadly diverse biological processes, including cell growth, differentiation, apoptosis, fetal development, transformation, inflammation, and immune response. Receptor-recruited STATs are phosphorylated on a single tyrosine residue in the carboxy terminal portion. The modified STATs are released from the cytoplasmic region of the receptor subunits to form homodimers or heterodimers through reciprocal interaction between the phosphotyrosine of one STAT and the SH2 domain of another. Following dimerization, STATs rapidly translocate to the nucleus and interact with specific regulatory elements to induce target gene transcription. Unlike all other members of the STAT gene family, ablation of STAT3 leads to embryonic lethality and it has been suggested that this protein might represent a primordial STAT protein. The function of Stat3 has been extensively studied in cell culture systems (reviewed in 3). It evokes a number of distinct responses in different cells, including induction of an acute-phase response in hepatoma cells, stimulation of proliferation in B lymphocytes, activation of terminal differentiation and growth arrest in monocytes, and maintenance of the pluripotency of embryonic stem cells. Dysregulation of STAT signaling pathways, particularly STAT3 and STAT5, has been demonstrated to contribute to malignant cellular transformation. Constitutive activation of STAT1, STAT3, and STAT5 is associated with malignant transformation induced by various oncoproteins. Src kinase-mediated activation of STAT3 has been shown to be essential in prostate and ovarian carcinomas. In head and neck cancers, constitutive STAT3 activity with up-regulated epidermal growth factor receptor (EGFR) signaling plays an important role in malignant proliferation. Altering the c-terminal domain of STAT3 induces constitutive activation and provides further evidence that STAT3 activation may be oncogenic by itself and is not just a consequence of tyrosine phosphorylation. Therefore, Stat3 may be a key player in the pathogenesis of diverse human cancers which makes this molecule a prime target for novel therapies.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeted disruption of the mouse Stat3 gene leads to early embryonic lethality.
Takeda K, et al.
Proceedings of the National Academy of Sciences of the USA, 94, 3801-3804 (1997)
YingYing Qian et al.
Molecular cancer, 13, 176-176 (2014-07-23)
Lung cancer in nonsmokers tends to be driven by a single somatic mutation or a gene fusion. KIF5B-RET fusion is an oncogene identified in non-small cell lung cancers. In this study, we verified the oncogenic activity of KIF5B-RET fusion and
STAT proteins and transcriptional responses to extracellular signals.
Horvath CM
Trends in Biochemical Sciences, 25, 496-502 (2000)
STATs and gene regulation.
Darnell JE Jr
Science, 277, 1630-1635 (1997)
Tracking STAT nuclear traffic.
Reich NC and Liu L
Nature Reviews: Immunology, 6, 602-612 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service