Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2012

Sigma-Aldrich

TFIID, native complex human

≥70% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

750 kDa

packaging

pkg of 2 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

50 μg/mL

color

clear colorless

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TAF2(6873)

Biochem/physiol Actions

TFIID is a multiprotein complex with a molecular mass of around 750 kDa that directs pre-initiation complex assembly on both TATA box-containing and TATA-less promoters. It consists of TATA-binding protein (TBP) and a number of TBP associated factors (TAFs). TBP alone can replace TFIID in a reconstituted in vitro transcription system but only the TFIID complex can mediate transcriptional activation. TAFII250, the largest subunit of TFIID contains protein kinase and histone acetyltransferase activities linking transcriptional initiation/activation with chromatin modification. In addition, multiple serine/ threonine phosphorylations of TBP and TAFs selectively inhibit the ability of TFIID to mediate transcriptional activation.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Matsui et al.
The Journal of biological chemistry, 255(24), 11992-11996 (1980-12-25)
A soluble extract prepared from human cells (KB S-100) has been recently shown to direct accurate transcription initiation by purified RNA polymerase II at the major late promoter of adenovirus 2. We have fractionated this extract by chromatography on phosphocellulose
B D Dynlacht et al.
Cell, 66(3), 563-576 (1991-08-09)
A key step in the regulation of transcription involves interactions between promoter-selective factors and various components of the transcriptional apparatus. Here we report the requirements for transcriptional activation directed by NTF-1, a developmentally regulated transcription factor in Drosophila. Reconstituted transcription
R Dikstein et al.
Cell, 84(5), 781-790 (1996-03-08)
Some TAF subunits of transcription factor TFIID play a pivotal role in transcriptional activation by mediating protein-protein interactions, whereas other TAFs direct promoter selectivity via protein-DNA recognition. Here, we report that purified recombinant TAFII250 is a protein serine kinase that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service