Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0732

Sigma-Aldrich

MEK5, Active

PRECISIO® Kinase, recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$456.00

$456.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
$456.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$456.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

70-100 nmol/min·mg (radiometric)

mol wt

~77 kDa

concentration

0.05-0.1 μg/μL

accession no.

BC008838

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MEK5(5607)

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Recombinant full-length human MEK5 was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. MEK5 or MAP2K5 (mitogen-activated protein kinase kinase 5) is a dual specificity protein kinase that belongs to the MAP kinase kinase family which specifically interacts with and activates MAPK7/ERK5. MEK5 itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by MEK5 is involved in growth factor stimulated cell proliferation and muscle cell differentiation (1). The rs3743354 polymorphism in the MEK5 promoter may affect the risk of developing colorectal cancer (CRC) (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service