Recombinant human TRKA (G667C) (440-end) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. TRKA is a member of the trk proto-oncogene family and encodes a 140kDa, membrane-spanning protein tyrosine kinase that is the functional receptor for nerve growth factor (NGF). NGF elicits the rapid phosphorylation of gp140trk on tyrosine residues leading to increased c-Fos expression, DNA synthesis and morphologic transformation (1). A decreased expression of TRKA on the striatal cholinergic neurons has been observed which may contribute, when it reaches a crucial threshold, to the death of cholinergic neurons observed in Alzheimer disease (2).
Reconstitution
Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.
Storage and Stability
Store in aliquots at -70°C. Avoid freeze/thaw cycles.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.