Recombinant full-length human ERK3 (MAPK6) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. ERK3 (MAPK6) or mitogen-activated protein kinase 6 is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases) which also known as extracellular signal-regulated kinases (ERKs) that are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals and tyrosine phosphorylated in response to insulin and NGF (1). ERK3 is highly expressed in various human tissues, most abundantly in skeletal muscle (2).
Reconstitution
Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.
Storage and Stability
Store in aliquots at -70°C. Avoid freeze/thaw cycles.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.