Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0719

Sigma-Aldrich

ALK2 (R206H), Active

PRECISIO® Kinase, recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$456.00

$456.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
$456.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$456.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

5-7 nmol/min·mg (radiometric)

mol wt

~67 kDa

concentration

0.1 μg/μL

accession no.

NM_001105

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ALK2(90)

Application

Useful for kinase assay.

Biochem/physiol Actions

Recombinant human ALK2 (R206H) (147-end) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. ALK2 belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins which encodes activin A type I receptor. ALK2 signals a particular transcriptional response in concert with activin type II receptors. Mutations in ALK2 are associated with fibrodysplasia ossificans progressiva (FOP) and the mutations are located in or adjacent to the GS regulatory region or active site of the kinase. The mutations in ALK2 were predicted by protein structure homology modeling to activate the ALK2 protein and enhance receptor signaling (1). Dysregulated ALK2 kinase activity plays a role in the pathogenesis of FOP (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service