Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0693

Sigma-Aldrich

PFKFB4 Protein

recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

recombinant

expressed in baculovirus infected Sf9 cells

assay

70-95% (SDS-PAGE)

form

frozen liquid

mol wt

~82 kDa

concentration

0.1 μg/μL

accession no.

NM_004567

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PFKFB4(5210)

Application

Useful for Western blot.

Biochem/physiol Actions

Recombinant human PFKFB4 (2-end) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. PFKFB4 (also known as 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4) regulates the steady-state concentration of fructose-2, 6-bisphosphate, a potent activator of a key regulatory enzyme of glycolysis pathway called phosphofructokinase. Isozymes of PFKFB differ in the regions surrounding the catalytic core, which are important for the differential response to allosteric effectors and hormonal signals in different tissues (1). PFKFB4 is highly expressed in brain, heart, liver, muscle, placenta, adipose tissue, ovary, fallopian tubes, and testis (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service