Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0685

Sigma-Aldrich

SAMSTIDE

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$105.00

$105.00


Usually ships in 2 weeks.


Select a Size

Change View
1 MG
$105.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$105.00


Usually ships in 2 weeks.

assay

≥95% (HPLC)

form

lyophilized powder

mol wt

1779.18

shipped in

ambient

storage temp.

−20°C

Application

Useful as a substrate for AMPK kinases.

Biochem/physiol Actions

The SAMStide peptide sequence (HMRSAMSGLHLVKRR) is based on the mouse acetyl-Coenzyme A carboxylase alpha (amino acid 73-85).

Sequence

HMRSAMSGLHLVKRR

Reconstitution

Reconstitute in 20mM Tris-HCl, pH 7.5 to a final concentration of 1mg/mL.

Storage and Stability

Store diluted product in aliquots at -20°C. Avoid freeze/thaw cycles.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service