Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0631

Sigma-Aldrich

PD-L1 (F19A), FC Fusion, Biotin Labeled

recombinant, expressed in HEK 293 cells

Synonym(s):

B7 homolog 1, B7-H1, CD274, PD-L1, PDCD1L1, Programmed Cell Death 1 Ligand 1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$471.00

$471.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 μG
$471.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$471.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

54 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD274(29126)

General description

Human secreted Programmed Cell Death 1 Ligand 1 (PD-L1)-Fc fusion protein, also known as CD274 and B7 homolog 1 (B7-H1) with a F19A mutation, GenBank Accession No. NM_014143,amino acids 19-239, fused at the Cterminus with the Fc portion of human IgG1,with C-terminal Avitag expressed in a HEK293 cell expression system and enzymatically biotin-labeled using Avitag technology. MW = 54 kDa.

Physical form

Aqueous solution

Legal Information

AviTag is a trademark of Avidity, LLC

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service