Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0486

Sigma-Aldrich

Bromodomain Non-Acetylated Ligand 1

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 ML
$761.00

$761.00


Usually ships in 2 weeks.


Select a Size

Change View
0.5 ML
$761.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$761.00


Usually ships in 2 weeks.

assay

≥95% (HPLC)

form

aqueous solution

mol wt

2602.1 kDa

packaging

pkg of 5 mL

shipped in

dry ice

storage temp.

−20°C

General description

The Bromodomain Non-acetylated Ligand 1 is peptide histone H4 (1-21). It contains an C-terminal GG linker, followed by a biotinylated Lys. Bromodomains function as acetyl-lysine binding domains, and bromodomain/acetyl-lysine recognition can regulate histone modification, DNA replications, chromatin remodeling, and gene transcription. This peptide is suitable for use as a ligand for several bromodomains including members of the BET bromodomain family: BRD2, BRD3, BRD4, and BRDT. It contains an C-terminal GG linker, followed by a biotinylated Lys. Bromodomains function as acetyl-lysine binding domains, and bromodomain/acetyl-lysine recognition can regulate histone modification, DNA replications, chromatin remodeling, and gene transcription. This peptide is suitable for use as a negative control for several bromodomains including ATAD2 and BAZ2B.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pil-Hoon Park et al.
American journal of physiology. Gastrointestinal and liver physiology, 289(6), G1124-G1136 (2005-08-06)
Ethanol treatment increases gene expression in the liver through mechanisms that are not clearly understood. Histone acetylation has been shown to induce transcriptional activation. We have investigated the characteristics and mechanisms of ethanol-induced histone H3 acetylation in rat hepatocytes. Immunocytochemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service