Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0477

Sigma-Aldrich

BRPF1 (627-746) human

recombinant, expressed in E. coli, ≥69% (SDS-PAGE)

Synonym(s):

BR140, bromodomain and PHD finger containing, 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$761.00

$761.00


Usually ships in 2 weeks.


Select a Size

Change View
100 μG
$761.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$761.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

assay

≥69% (SDS-PAGE)

form

aqueous solution

mol wt

14.3 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRPF1(7862)

General description

Human Bromodomain and PHD finger containing 1, or BRPF1 (GenBank Accession No. NM_001003694) amino-acids 627 – 746 with N-terminal HIS-tag, MW = 15.1 kDa, expressed in an E. coli expression system.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laue, K., et al.
Development, 1935-1946 (2008)
Viveka Mayya et al.
Science signaling, 2(84), ra46-ra46 (2009-08-20)
Protein phosphorylation events during T cell receptor (TCR) signaling control the formation of complexes among proteins proximal to the TCR, the activation of kinase cascades, and the activation of transcription factors; however, the mode and extent of the influence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service