Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0443

Sigma-Aldrich

UCHL3 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Synonym(s):

UCHL3, Ubiquitin Carboxy-terminal Hydrolase L3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

27.1 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

2.07 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... UCHL3(7347)

General description

UCHL3, also known as Ubiquitin Carboxy-terminal Hydrolase L3 (GenBank Accession No. NM_006002) amino acids 2-231 (end) with N-terminal His-tag, MW=27.1 kDa, expressed in E. coli.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nam, M.J., et al.
Proteomics, 3, 2108-2115 (2004)
H Wada et al.
Biochemical and biophysical research communications, 251(3), 688-692 (1998-10-29)
NEDD8 is a novel ubiquitin-like protein that has been shown to conjugate to nuclear proteins in a manner analogous to ubiquitination and sentrinization. To identify proteins that are involved in the NEDD8-conjugation and de-conjugation pathway, the yeast two-hybrid system was
Rieko Setsuie et al.
Neurochemistry international, 54(5-6), 314-321 (2009-01-22)
Ubiquitin (Ub) carboxy terminal hydrolase (UCH)-L1 and UCH-L3 are two of the deubiquitinating enzymes expressed in the brain. Both gad mice, which lack UCH-L1 expression and Uchl3 knockout mice exhibit neurodegeneration, although at distinct areas. These phenotypes indicate the importance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service