Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0431

Sigma-Aldrich

PDE4D7 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥65% (SDS-PAGE)

Synonym(s):

DPDE3, PDE43, STRK1, phosphodiesterase 4D, cAMP-specific

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥65% (SDS-PAGE)

form

aqueous solution

mol wt

111 kDa

packaging

pkg of 5 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDE4D(5144)

General description

Human PDE4D7 (GenBank Accession No. NM_001165899) amino acids 2-748 (end) with N-terminal GST-tag, MW=111 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miles D Houslay
Molecular pharmacology, 68(3), 563-567 (2005-06-17)
In this issue, Tilley and Maurice (p. 596) show that differentiation of vascular smooth muscle cells to a proliferative phenotype is associated with a profound up-regulation of specific phosphodiesterase-4 (PDE4) isoforms because of increased histone acetylation. The increased PDE4 activity
Daguang Wang et al.
Cellular signalling, 15(9), 883-891 (2003-07-02)
We report here the cloning and characterization of two novel PDE4D isoforms, PDE4D6 and PDE4D7. PDE4D6 is a supershort form and PDE4D7 a long form of PDE4D. In addition, we have identified another novel long-form variant, PDE4D8, in silico. Like

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service